Learn More
Waters Corp Sep-Pak Vac Cartridge, C18, 200 mg, 55 – 105 μm
Catalog No. 50785823 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Vac Cartridge is a powerful tool for solid phase extraction, featuring a silica-based octadecyl bonded phase with strong hydrophobicity. It is designed for use with vacuum manifolds and automated SPE instruments, adsorbing even weakly hydrophobic analytes from aqueous solutions. It is useful for analyzing drugs and metabolites, desalting peptides, and detecting trace organics in environmental water.
  • Possesses strong hydrophobicity, suitable for weakly hydrophobic analytes.
  • Designed for vacuum manifolds and automated solid phase extraction instruments.
  • Assures absorbent quality, reproducibility, versatility, and ease of use.
  • Manufactured under ISO 9002, GLP, and cGMP guidelines.
  • Ensures consistent solid phase chemistry across batches.
  • Provides lower costs, improved recoveries, and faster protocols.
  • Enhances safety and prevents cross-contamination.
Catalog No. 50-785-823 Supplier Waters Corp Supplier No. WAT054945
This Vac Cartridge is a powerful tool for solid phase extraction, featuring a silica-based octadecyl bonded phase with strong hydrophobicity. It is designed for use with vacuum manifolds and automated SPE instruments, adsorbing even weakly hydrophobic analytes from aqueous solutions. It is useful for analyzing drugs and metabolites, desalting peptides, and detecting trace organics in environmental water.
  • Possesses strong hydrophobicity, suitable for weakly hydrophobic analytes.
  • Designed for vacuum manifolds and automated solid phase extraction instruments.
  • Assures absorbent quality, reproducibility, versatility, and ease of use.
  • Manufactured under ISO 9002, GLP, and cGMP guidelines.
  • Ensures consistent solid phase chemistry across batches.
  • Provides lower costs, improved recoveries, and faster protocols.
  • Enhances safety and prevents cross-contamination.