Learn More
Waters Corp Sep-Pak Vac Cartridge, C18, 6 cc, 1 g Sorbent, 55 to 105 μm
Catalog No. 50785407 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Vac Cartridge features a silica-based octadecyl bonded phase with strong hydrophobicity, designed to adsorb analytes from aqueous solutions. Typical applications include drugs and metabolites in biological fluids, desalting peptides, and analyzing trace organics in environmental water.
  • Designed for sample enrichment and purification.
  • Contains high-quality, reproducible packings.
  • Offers optimal surface area, pore-size, and particle-size distributions.
  • Sealed in a non-contaminating poly-foil pouch for product integrity.
  • Compatible with vacuum manifolds for steady extraction and filtration and processing multiple samples simultaneously.
Catalog No. 50-785-407 Supplier Waters Corp Supplier No. WAT036905
This Vac Cartridge features a silica-based octadecyl bonded phase with strong hydrophobicity, designed to adsorb analytes from aqueous solutions. Typical applications include drugs and metabolites in biological fluids, desalting peptides, and analyzing trace organics in environmental water.
  • Designed for sample enrichment and purification.
  • Contains high-quality, reproducible packings.
  • Offers optimal surface area, pore-size, and particle-size distributions.
  • Sealed in a non-contaminating poly-foil pouch for product integrity.
  • Compatible with vacuum manifolds for steady extraction and filtration and processing multiple samples simultaneously.