Learn More
Waters Corp Waters Corporation Sep-Pak Vac Cartridge, C18, 6 cc
Catalog No. 50785510 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based octadecyl bonded phase vac cartridge is designed for solid phase extraction. It effectively adsorbs analytes, even those with weak hydrophobicity, from aqueous solutions. Common applications include analysis of drugs and metabolites, desalting peptides, and screening environmental water samples.
  • Highly referenced solid phase extraction product for sample preparation.
  • Suitable for various analysis methods, including GC, HPLC, and UPLC.
  • Manufactured in an ISO 13485 and ISO 9002 registered facility, compliant with cGMP guidelines for US FDA Class 1 Medical Devices.
  • Features C18 chemistry and is endcapped.
  • Reversed phase mode for efficient separation.
  • Particle size range of 55–105 μm.
  • Pore size of 125 Å.
  • Sorbent weight of 500 mg.
  • Barrel size of 6 cc with a hold up volume of 1.2 mL.
  • Operates within a pH range of 2 to 8.
Catalog No. 50-785-510 Supplier Waters Corp Supplier No. WAT043395
This silica-based octadecyl bonded phase vac cartridge is designed for solid phase extraction. It effectively adsorbs analytes, even those with weak hydrophobicity, from aqueous solutions. Common applications include analysis of drugs and metabolites, desalting peptides, and screening environmental water samples.
  • Highly referenced solid phase extraction product for sample preparation.
  • Suitable for various analysis methods, including GC, HPLC, and UPLC.
  • Manufactured in an ISO 13485 and ISO 9002 registered facility, compliant with cGMP guidelines for US FDA Class 1 Medical Devices.
  • Features C18 chemistry and is endcapped.
  • Reversed phase mode for efficient separation.
  • Particle size range of 55–105 μm.
  • Pore size of 125 Å.
  • Sorbent weight of 500 mg.
  • Barrel size of 6 cc with a hold up volume of 1.2 mL.
  • Operates within a pH range of 2 to 8.