Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, C8, 12 cc, 2 g Sorbent
Catalog No. 50785764 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Vac cartridge features a silica-based bonded phase with moderate hydrophobicity. It is designed for methods requiring less retention than C18. Typical applications include the analysis of drugs and their metabolites in serum, plasma or urine, and peptides in serum, plasma.
  • Silica-based bonded phase with moderate hydrophobicity
  • Suitable for applications requiring less retention than C18
  • Mass spec compatibility: Yes
  • Endcapped: Yes
  • Mode: Reversed phase
  • Water wettable: No
Catalog No. 50-785-764 Supplier Waters Corp Supplier No. WAT054615
This Vac cartridge features a silica-based bonded phase with moderate hydrophobicity. It is designed for methods requiring less retention than C18. Typical applications include the analysis of drugs and their metabolites in serum, plasma or urine, and peptides in serum, plasma.
  • Silica-based bonded phase with moderate hydrophobicity
  • Suitable for applications requiring less retention than C18
  • Mass spec compatibility: Yes
  • Endcapped: Yes
  • Mode: Reversed phase
  • Water wettable: No