Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Solid Phase Extraction Cartridge, C8, 1 cc, 100 mg, 37–55 μm

Catalog No. 50785394 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This silica-based bonded phase cartridge offers moderate hydrophobicity, suitable for methods requiring less retention than C18. It is typically used for applications such as analyzing drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma.

  • Chemistry: C8
  • Endcapped: Yes
  • Format: Vac Cartridge
  • Mass spec compatibility: Yes
  • Mode: Reversed phase
  • Particle size: 37 to 55 μm
  • Pore size: 125 Å
  • Sorbent substrate: Silica
  • Sorbent weight: 100 mg
  • Water wettable: No
  • pH range: 2 to 8 pH
  • Barrel size: 1 cc
  • Hold up volume: 0.25 mL

Catalog No. 50-785-394 Supplier Waters Corp Supplier No. WAT036785
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This silica-based bonded phase cartridge offers moderate hydrophobicity, suitable for methods requiring less retention than C18. It is typically used for applications such as analyzing drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma.

  • Chemistry: C8
  • Endcapped: Yes
  • Format: Vac Cartridge
  • Mass spec compatibility: Yes
  • Mode: Reversed phase
  • Particle size: 37 to 55 μm
  • Pore size: 125 Å
  • Sorbent substrate: Silica
  • Sorbent weight: 100 mg
  • Water wettable: No
  • pH range: 2 to 8 pH
  • Barrel size: 1 cc
  • Hold up volume: 0.25 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.