Learn More
Waters Corp Sep-Pak Vac Cartridge, C8, 1 cc, 50 mg sorbent
Catalog No. 50785826 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Vac Cartridge features a silica-based bonded phase with moderate hydrophobicity, designed for methods requiring less retention than C18. It is typically used for the analysis of drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma. Each cartridge contains 50 mg of sorbent and is sold in packages of 100 pieces.
  • Used for methods requiring less retention than C18.
  • Suitable for drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma.
  • Manufactured to ISO 9001, ISO 13485, and cGMP standards.
  • Simple to use, reproducible, and disposable.
  • Packings are clean, dry, and reproducible in activity and capacity.
Catalog No. 50-785-826 Supplier Waters Corp Supplier No. WAT054965
This Vac Cartridge features a silica-based bonded phase with moderate hydrophobicity, designed for methods requiring less retention than C18. It is typically used for the analysis of drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma. Each cartridge contains 50 mg of sorbent and is sold in packages of 100 pieces.
  • Used for methods requiring less retention than C18.
  • Suitable for drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma.
  • Manufactured to ISO 9001, ISO 13485, and cGMP standards.
  • Simple to use, reproducible, and disposable.
  • Packings are clean, dry, and reproducible in activity and capacity.