Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Vac Cartridge, C8, 20 cc, 5 g Sorbent, 37 – 55 μm

Catalog No. 50785773 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This cartridge features a silica-based bonded phase with moderate hydrophobicity, making it suitable for methods requiring less retention compared to C18 chemistries. Typical applications include the extraction of drugs and their metabolites from serum, plasma, or urine, as well as peptides from serum and plasma.

  • Chemistry: C8
  • Endcapped: Yes
  • Format: Vac Cartridge
  • Mass spec compatibility: Yes
  • Mode: Reversed phase
  • Particle size: 37 – 55 μm
  • Pore size: 125 Å
  • Sorbent substrate: Silica
  • Sorbent weight: 5 g
  • Water wettable: No
  • pH range: 2 to 8
  • Barrel size: 20 cc
  • Hold up volume: 7.8 mL

Catalog No. 50-785-773 Supplier Waters Corp Supplier No. WAT054660
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This cartridge features a silica-based bonded phase with moderate hydrophobicity, making it suitable for methods requiring less retention compared to C18 chemistries. Typical applications include the extraction of drugs and their metabolites from serum, plasma, or urine, as well as peptides from serum and plasma.

  • Chemistry: C8
  • Endcapped: Yes
  • Format: Vac Cartridge
  • Mass spec compatibility: Yes
  • Mode: Reversed phase
  • Particle size: 37 – 55 μm
  • Pore size: 125 Å
  • Sorbent substrate: Silica
  • Sorbent weight: 5 g
  • Water wettable: No
  • pH range: 2 to 8
  • Barrel size: 20 cc
  • Hold up volume: 7.8 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.