Learn More
Waters Corp Sep-Pak C8 35 cc Vac Cartridge, 10 g Sorbent per Cartridge, 37 - 55 µm, 10/pk
Catalog No. 50785781 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Silica-based bonded phase with moderate hydrophobicity; use for methods requiring less retention than C18; typical applications include drugs and their metabolites in serum, plasma or urine, peptides in serum, plasma.
Catalog No. 50-785-781 Supplier Waters Corp Supplier No. WAT054700
Silica-based bonded phase with moderate hydrophobicity; use for methods requiring less retention than C18; typical applications include drugs and their metabolites in serum, plasma or urine, peptides in serum, plasma.