Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Vac Cartridge, C8, 200 mg Sorbent, 37 to 55 μm

Catalog No. 50785822 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Vac Cartridge features a silica-based bonded phase with moderate hydrophobicity. It is designed for methods requiring less retention than C18 and is typically used for applications such as analyzing drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma.

  • C8 chemistry
  • Endcapped
  • Vac cartridge format
  • Mass spec compatible
  • Reversed phase mode
  • Not water wettable
  • Minimum pH range: 2 pH
  • Maximum pH range: 8 pH
  • Cartridge volume: 3 cc
  • Sorbent weight: 200 mg
  • Particle size: 37 to 55 μm
  • Pack quantity: 50 pk
  • Silica sorbent substrate
  • Pore size: 125 Å
  • Hold up volume: 0.34 mL

Catalog No. 50-785-822 Supplier Waters Corp Supplier No. WAT054940
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Vac Cartridge features a silica-based bonded phase with moderate hydrophobicity. It is designed for methods requiring less retention than C18 and is typically used for applications such as analyzing drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma.

  • C8 chemistry
  • Endcapped
  • Vac cartridge format
  • Mass spec compatible
  • Reversed phase mode
  • Not water wettable
  • Minimum pH range: 2 pH
  • Maximum pH range: 8 pH
  • Cartridge volume: 3 cc
  • Sorbent weight: 200 mg
  • Particle size: 37 to 55 μm
  • Pack quantity: 50 pk
  • Silica sorbent substrate
  • Pore size: 125 Å
  • Hold up volume: 0.34 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.