Learn More
Waters Corp Waters SPE Cartridge, C8, 3 cc, 500 mg, 37-55 μm
Catalog No. 50785393 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based, endcapped, reversed-phase solid phase extraction device is designed for methods requiring moderate hydrophobicity and less retention than C18. It features 500 mg of sorbent per cartridge, a particle size of 37 to 55 μm, and a 3 cc barrel size, suitable for analyzing drugs, metabolites, and peptides in biological matrices.
  • Silica-based bonded phase with moderate hydrophobicity.
  • Designed for methods requiring less retention than C18.
  • Optimized packings for tidy, dry, and repeatable sample preparation; ideal surface area, pore size, and particle size distributions.
  • Product integrity maintained by non-contaminating poly-foil packaging.
  • Frit from HD and UHMW polyethylenes, offering excellent solvent resistance, low extractables, and depth filtration.
  • Suitable for analysis of drugs, metabolites, and peptides in biological matrices.
Catalog No. 50-785-393 Supplier Waters Corp Supplier No. WAT036780
This silica-based, endcapped, reversed-phase solid phase extraction device is designed for methods requiring moderate hydrophobicity and less retention than C18. It features 500 mg of sorbent per cartridge, a particle size of 37 to 55 μm, and a 3 cc barrel size, suitable for analyzing drugs, metabolites, and peptides in biological matrices.
  • Silica-based bonded phase with moderate hydrophobicity.
  • Designed for methods requiring less retention than C18.
  • Optimized packings for tidy, dry, and repeatable sample preparation; ideal surface area, pore size, and particle size distributions.
  • Product integrity maintained by non-contaminating poly-foil packaging.
  • Frit from HD and UHMW polyethylenes, offering excellent solvent resistance, low extractables, and depth filtration.
  • Suitable for analysis of drugs, metabolites, and peptides in biological matrices.