Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Sep-Pak Vac Cartridge, C8, 6 cc, 500 mg, 37-55 μm

Catalog No. 50785746 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This silica-based bonded phase product offers moderate hydrophobicity, designed for methods requiring less retention than C18. It is suitable for applications such as the analysis of drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma.

  • Sorbent type: C8, reversed phase, silica
  • Cartridge size: 6 cc
  • Sorbent mass: 500 mg
  • Particle size: 37 to 55 μm
  • Quantity per pack: 30 pk
  • Endcapped: Yes
  • Format: Vac Cartridge
  • Mass spec compatibility: Yes
  • Pore size: 125 Å
  • Water wettable: No
  • pH range: 2 to 8
  • Hold up volume: 1.1 mL

Catalog No. 50-785-746 Supplier Waters Corp Supplier No. WAT054525
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This silica-based bonded phase product offers moderate hydrophobicity, designed for methods requiring less retention than C18. It is suitable for applications such as the analysis of drugs and their metabolites in serum, plasma, or urine, and peptides in serum or plasma.

  • Sorbent type: C8, reversed phase, silica
  • Cartridge size: 6 cc
  • Sorbent mass: 500 mg
  • Particle size: 37 to 55 μm
  • Quantity per pack: 30 pk
  • Endcapped: Yes
  • Format: Vac Cartridge
  • Mass spec compatibility: Yes
  • Pore size: 125 Å
  • Water wettable: No
  • pH range: 2 to 8
  • Hold up volume: 1.1 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.