Learn More
Waters Corp Sep-Pak Vac Cartridge, C8, 6 cc, 500 mg
Catalog No. 50785755 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid-phase extraction (SPE) cartridge is designed for sample preparation in various analytical methods such as GC, HPLC, and UPLC. It features a C8 sorbent, a 6 cc cartridge volume, and 500 mg of sorbent material.
  • C8 sorbent type.
  • 6 cc cartridge volume.
  • 500 mg sorbent weight.
  • Particle size of 55–105 μm.
  • Suitable for sample preparation in GC, HPLC, and UPLC analysis methods.
Catalog No. 50-785-755 Supplier Waters Corp Supplier No. WAT054570
This solid-phase extraction (SPE) cartridge is designed for sample preparation in various analytical methods such as GC, HPLC, and UPLC. It features a C8 sorbent, a 6 cc cartridge volume, and 500 mg of sorbent material.
  • C8 sorbent type.
  • 6 cc cartridge volume.
  • 500 mg sorbent weight.
  • Particle size of 55–105 μm.
  • Suitable for sample preparation in GC, HPLC, and UPLC analysis methods.