Learn More
Waters Corp Sep-Pak Cyanopropyl (CN) 20 cc Vac Cartridge, 5 g Sorbent per Cartridge, 55 - 105 µm, 20/pk
Catalog No. 50785778 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Silica-based polar bonded phase; can be used as less polar alternative to silica in normal-phase applications or as less hydrophobic alternative to C18 or C8 in reversed-phase applications; typical applications include drug metabolites and pesticides.
Catalog No. 50-785-778 Supplier Waters Corp Supplier No. WAT054685
Silica-based polar bonded phase; can be used as less polar alternative to silica in normal-phase applications or as less hydrophobic alternative to C18 or C8 in reversed-phase applications; typical applications include drug metabolites and pesticides.