Learn More
Waters Corp WATERS Sep-Pak Vac Cartridge, Cyanopropyl (CN), 500 mg, 3 cc
Catalog No. 50785116 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Vac Cartridge features a silica-based polar bonded phase. It can be used as a less polar alternative to silica in normal-phase applications or as a less hydrophobic alternative to C18 or C8 in reversed-phase applications. Typical applications include drug metabolites and pesticides.
  • Silica-based polar bonded phase
  • Functions as a less polar alternative to silica in normal-phase applications
  • Functions as a less hydrophobic alternative to C18 or C8 in reversed-phase applications
  • Suitable for applications including drug metabolites and pesticides
  • Endcapped for enhanced performance
  • Mass spectrometry compatible
  • Operates within a pH range of 2 to 8
  • Has a hold up volume of 0.8 mL
  • Features a particle size of 55 to 105 μm
  • Sorbent weight of 500 mg
  • Pore size of 125 Å
Catalog No. 50-785-116 Supplier Waters Corp Supplier No. WAT020835
This Vac Cartridge features a silica-based polar bonded phase. It can be used as a less polar alternative to silica in normal-phase applications or as a less hydrophobic alternative to C18 or C8 in reversed-phase applications. Typical applications include drug metabolites and pesticides.
  • Silica-based polar bonded phase
  • Functions as a less polar alternative to silica in normal-phase applications
  • Functions as a less hydrophobic alternative to C18 or C8 in reversed-phase applications
  • Suitable for applications including drug metabolites and pesticides
  • Endcapped for enhanced performance
  • Mass spectrometry compatible
  • Operates within a pH range of 2 to 8
  • Has a hold up volume of 0.8 mL
  • Features a particle size of 55 to 105 μm
  • Sorbent weight of 500 mg
  • Pore size of 125 Å