Learn More
Waters Corp Waters Sep-Pak Cartridge, Cyanopropyl (CN), Vac 3 cc/200 mg, 50/box
Catalog No. 50785821 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Cyanopropyl (CN) Sep-Pak cartridge is a silica-based Solid-Phase Extraction (SPE) product, known for pioneering sample preparation and being widely referenced. These cartridges are precision chromatographic tools for sample enrichment and purification, offering a 3 mL volume and 200 mg of Cyanopropyl (CN) sorbent, with a hold up volume of 0.42 mL, provided in a 50-count box.
  • Offers minimum cost sample preparation.
  • Constructed with molded, high purity polypropylene “syringe-barrel” bodies.
  • Includes two polyethylene frits to contain the packing material.
  • Features a standard male-Luer tip for use with multi-position vacuum manifolds.
  • Color-coded printing on the syringe barrels identifies the sorbent type.
Catalog No. 50-785-821 Supplier Waters Corp Supplier No. WAT054935
The Cyanopropyl (CN) Sep-Pak cartridge is a silica-based Solid-Phase Extraction (SPE) product, known for pioneering sample preparation and being widely referenced. These cartridges are precision chromatographic tools for sample enrichment and purification, offering a 3 mL volume and 200 mg of Cyanopropyl (CN) sorbent, with a hold up volume of 0.42 mL, provided in a 50-count box.
  • Offers minimum cost sample preparation.
  • Constructed with molded, high purity polypropylene “syringe-barrel” bodies.
  • Includes two polyethylene frits to contain the packing material.
  • Features a standard male-Luer tip for use with multi-position vacuum manifolds.
  • Color-coded printing on the syringe barrels identifies the sorbent type.