Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Sep-Pak Vac Cartridge, Diol, 12 cc, 2 g Sorbent

Catalog No. 50785772 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This product is a silica-based polar bonded phase with neutral character. It can be used as an alternative to silica in normal-phase applications where the acidic character of silica is undesirable, or as a very weakly interacting phase in aqueous applications. Its applications include the extraction of antibiotics from cosmetics and the isolation of proteins or peptides by hydrophobic interaction chromatography.

  • Solid phase extraction product
  • 20 units per package
  • Diol chemistry
  • Not endcapped
  • Vac cartridge format
  • Mass spec compatible
  • Reversed phase mode
  • Particle size: 37 to 55 μm
  • Pore size: 300 Å
  • Silica sorbent substrate
  • Sorbent weight: 2 g
  • Not water wettable
  • pH range: 2 to 8
  • Barrel size: 12 cc
  • Hold up volume: 3.5 mL

Catalog No. 50-785-772 Supplier Waters Corp Supplier No. WAT054655
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a silica-based polar bonded phase with neutral character. It can be used as an alternative to silica in normal-phase applications where the acidic character of silica is undesirable, or as a very weakly interacting phase in aqueous applications. Its applications include the extraction of antibiotics from cosmetics and the isolation of proteins or peptides by hydrophobic interaction chromatography.

  • Solid phase extraction product
  • 20 units per package
  • Diol chemistry
  • Not endcapped
  • Vac cartridge format
  • Mass spec compatible
  • Reversed phase mode
  • Particle size: 37 to 55 μm
  • Pore size: 300 Å
  • Silica sorbent substrate
  • Sorbent weight: 2 g
  • Not water wettable
  • pH range: 2 to 8
  • Barrel size: 12 cc
  • Hold up volume: 3.5 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.