Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Diol, 100 mg Sorbent, 37 to 55 μm, 1 cc
Catalog No. 50785192 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based polar bonded phase cartridge offers a neutral character. It can be used in normal-phase applications where acidic silica is undesirable, or as a very weakly interacting phase in aqueous applications. It is suitable for isolating antibiotics from cosmetics and proteins or peptides via hydrophobic interaction chromatography.
  • Diol chemistry provides a polar bonded phase with neutral character.
  • Can be used in normal-phase applications where acidic silica is not desired.
  • Suitable for aqueous applications as a very weakly interacting phase.
  • Features a particle size of 37 to 55 μm.
  • Has a pore size of 300 Å.
  • Contains 100 mg of sorbent.
  • Designed with a 1 mL barrel size.
  • Hold up volume: 0.25 mL.
  • Mass spectrometry compatible.
  • Operates within a pH range of 2 to 8.
  • Not endcapped.
  • Not water wettable.
Catalog No. 50-785-192 Supplier Waters Corp Supplier No. WAT023605
This silica-based polar bonded phase cartridge offers a neutral character. It can be used in normal-phase applications where acidic silica is undesirable, or as a very weakly interacting phase in aqueous applications. It is suitable for isolating antibiotics from cosmetics and proteins or peptides via hydrophobic interaction chromatography.
  • Diol chemistry provides a polar bonded phase with neutral character.
  • Can be used in normal-phase applications where acidic silica is not desired.
  • Suitable for aqueous applications as a very weakly interacting phase.
  • Features a particle size of 37 to 55 μm.
  • Has a pore size of 300 Å.
  • Contains 100 mg of sorbent.
  • Designed with a 1 mL barrel size.
  • Hold up volume: 0.25 mL.
  • Mass spectrometry compatible.
  • Operates within a pH range of 2 to 8.
  • Not endcapped.
  • Not water wettable.