Learn More
Waters Corp Waters Corporation Sep-Pak Vac Cartridge, 3 cc, 500 mg, 37–55 μm, Diol
Catalog No. 50785118 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a silica-based polar bonded phase with a neutral character. It can serve as an alternative to silica in normal-phase applications where the acidic nature of silica is not desired, or as a very weakly interacting phase in aqueous applications.
  • Silica-based polar bonded phase with neutral character.
  • Can be used as an alternative to silica in normal-phase applications where the acidic character of silica is undesirable.
  • Can be used as a very weakly interacting phase in aqueous applications.
  • Suitable for applications including isolating antibiotics from cosmetics.
  • Suitable for isolating proteins or peptides via hydrophobic interaction chromatography.
Catalog No. 50-785-118 Supplier Waters Corp Supplier No. WAT020845
This is a silica-based polar bonded phase with a neutral character. It can serve as an alternative to silica in normal-phase applications where the acidic nature of silica is not desired, or as a very weakly interacting phase in aqueous applications.
  • Silica-based polar bonded phase with neutral character.
  • Can be used as an alternative to silica in normal-phase applications where the acidic character of silica is undesirable.
  • Can be used as a very weakly interacting phase in aqueous applications.
  • Suitable for applications including isolating antibiotics from cosmetics.
  • Suitable for isolating proteins or peptides via hydrophobic interaction chromatography.