Learn More
Waters Corp Waters Sep-Pak Diol Vac Cartridge, 6 cc, 500 mg, 37–55 μm
Catalog No. 50785754 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based polar bonded phase, with neutral character, serves as an alternative to silica in normal-phase applications where acidic characteristics are undesirable. It is also suitable as a very weakly interacting phase in aqueous applications, engineered for clean, dry, and reproducible activity and capacity.
  • High-quality sorbent design tailored for sample processing.
  • Tested for chromatographic activity, retention, and selectivity with each lot.
  • Packaged in a polyfoil pouch to protect integrity, inertness, and purity.
  • Optimized packings for sample preparation, ensuring cleanliness, dryness, and consistent activity and capacity.
  • Facilitates quick and easy extraction of solvents, solutions, and samples.
  • Allows transfer without spillage or sample loss due to evaporation.
  • Precision chromatographic device for sample enrichment and purification.
Catalog No. 50-785-754 Supplier Waters Corp Supplier No. WAT054565
This silica-based polar bonded phase, with neutral character, serves as an alternative to silica in normal-phase applications where acidic characteristics are undesirable. It is also suitable as a very weakly interacting phase in aqueous applications, engineered for clean, dry, and reproducible activity and capacity.
  • High-quality sorbent design tailored for sample processing.
  • Tested for chromatographic activity, retention, and selectivity with each lot.
  • Packaged in a polyfoil pouch to protect integrity, inertness, and purity.
  • Optimized packings for sample preparation, ensuring cleanliness, dryness, and consistent activity and capacity.
  • Facilitates quick and easy extraction of solvents, solutions, and samples.
  • Allows transfer without spillage or sample loss due to evaporation.
  • Precision chromatographic device for sample enrichment and purification.