Learn More
Waters Corp Sep-Pak Diol 6 cc Vac Cartridge, 1 g Sorbent per Cartridge, 37 - 55 µm, 30/pk
Catalog No. 50785763 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Silica-based polar bonded phase with neutral character; can be used as an alternative to silica in normal-phase applications, where the acidic character of silica is undesirable or as very weakly interacting phase in aqueous applications; applications include antibiotics from cosmetics; isolation of proteins or peptides by hydrophobic interaction chromatography.
Catalog No. 50-785-763 Supplier Waters Corp Supplier No. WAT054610
Silica-based polar bonded phase with neutral character; can be used as an alternative to silica in normal-phase applications, where the acidic character of silica is undesirable or as very weakly interacting phase in aqueous applications; applications include antibiotics from cosmetics; isolation of proteins or peptides by hydrophobic interaction chromatography.