Learn More
Waters Corp Sep-Pak Vac Cartridge, Florisil, 500 mg, 6 cc, 50–200 μm
Catalog No. 50785512 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This vac cartridge features Florisil, an extremely polar sorbent (magnesium oxide and silica gel blend) for adsorbing low to moderate polarity species. It's used for pesticide analysis in regulated food and environmental methods. Designed for sample enrichment and purification, it aids in easy extraction and transfer without spillage or evaporation loss. Product integrity is maintained by a non-contaminating poly-foil pouch.
  • Highly active, weakly basic sorbent.
  • Effective for pesticide analysis.
  • Facilitates convenient sample transfer.
  • High-quality, reproducible packings.
  • 20 μm frit offers solvent resistance and good flow.
  • Frit acts as a depth filter.
Catalog No. 50-785-512 Supplier Waters Corp Supplier No. WAT043405
This vac cartridge features Florisil, an extremely polar sorbent (magnesium oxide and silica gel blend) for adsorbing low to moderate polarity species. It's used for pesticide analysis in regulated food and environmental methods. Designed for sample enrichment and purification, it aids in easy extraction and transfer without spillage or evaporation loss. Product integrity is maintained by a non-contaminating poly-foil pouch.
  • Highly active, weakly basic sorbent.
  • Effective for pesticide analysis.
  • Facilitates convenient sample transfer.
  • High-quality, reproducible packings.
  • 20 μm frit offers solvent resistance and good flow.
  • Frit acts as a depth filter.