Learn More
Waters Corp Waters Vac Cartridge, Florisil, 3 cc, 500 mg, 50 to 200 μm
Catalog No. 50785112 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Vac Cartridge utilizes Florisil™, an extremely polar sorbent made from a blend of magnesium oxide and silica gel. Designed for pesticide analysis in food and environmental samples, this highly active and weakly basic sorbent adsorbs low to moderate polarity species from nonaqueous solutions. It serves as a precision chromatographic tool for sample enrichment and purification.
  • Sealed in a non-contaminating poly-foil pouch to preserve product integrity, packing inactivity, and purity.
  • Contains high-quality packing materials, specifically created for sample processing, which are clean, dry, and provide reproducible activity and capacity.
  • Equipped with a 20 μm frit made of a blend of HD and UHMW polyethylene, offering high solvent resistance, extremely low extractable levels, strong flow characteristics, and acting as a depth filter to capture minute amounts of sample detritus.
Catalog No. 50-785-112 Supplier Waters Corp Supplier No. WAT020815
This Vac Cartridge utilizes Florisil™, an extremely polar sorbent made from a blend of magnesium oxide and silica gel. Designed for pesticide analysis in food and environmental samples, this highly active and weakly basic sorbent adsorbs low to moderate polarity species from nonaqueous solutions. It serves as a precision chromatographic tool for sample enrichment and purification.
  • Sealed in a non-contaminating poly-foil pouch to preserve product integrity, packing inactivity, and purity.
  • Contains high-quality packing materials, specifically created for sample processing, which are clean, dry, and provide reproducible activity and capacity.
  • Equipped with a 20 μm frit made of a blend of HD and UHMW polyethylene, offering high solvent resistance, extremely low extractable levels, strong flow characteristics, and acting as a depth filter to capture minute amounts of sample detritus.