Learn More
Waters Corp Waters Vac Cartridge, Florisil, 12 cc, 2 g Sorbent
Catalog No. 50785508 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak Florisil 12 cc Vac Cartridge is a precision chromatographic instrument for sample enrichment and purification, extracting solvents and samples without spillage or loss. Florisil™, a polar, active, and weakly basic sorbent from magnesium oxide and silica gel, is ideal for adsorbing low to moderate polarity species from non-aqueous solutions. Cartridges are sealed in a non-contaminating poly-foil pouch with high-quality, clean, dry, and consistent packings.
  • Polar, highly active, weakly basic sorbent
  • Adsorbs low to moderate polarity species from nonaqueous solutions
  • Used for pesticide analysis in food and environmental samples via AOAC, EPA, and JPMHLW regulatory techniques
  • Sealed in a non-contaminating poly-foil pouch
  • High-quality, clean, dry, and repeatable packings
  • Water wettable
  • Mass spec compatibility: yes
  • Mode: normal-phase
Catalog No. 50-785-508 Supplier Waters Corp Supplier No. WAT043385
The Sep-Pak Florisil 12 cc Vac Cartridge is a precision chromatographic instrument for sample enrichment and purification, extracting solvents and samples without spillage or loss. Florisil™, a polar, active, and weakly basic sorbent from magnesium oxide and silica gel, is ideal for adsorbing low to moderate polarity species from non-aqueous solutions. Cartridges are sealed in a non-contaminating poly-foil pouch with high-quality, clean, dry, and consistent packings.
  • Polar, highly active, weakly basic sorbent
  • Adsorbs low to moderate polarity species from nonaqueous solutions
  • Used for pesticide analysis in food and environmental samples via AOAC, EPA, and JPMHLW regulatory techniques
  • Sealed in a non-contaminating poly-foil pouch
  • High-quality, clean, dry, and repeatable packings
  • Water wettable
  • Mass spec compatibility: yes
  • Mode: normal-phase