Learn More
Waters Corp Sep-Pak Vac Cartridge, Aminopropyl (NH2), 12 cc, 2 g
Catalog No. 50785771 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Amino Propyl (NH2) Vac cartridges contain a moderately polar, silica-based bonded phase with a weakly basic surface.
  • Sorbent can function as a polar sorbent, offering different selectivity for acidic/basic analytes, or as a weak anion exchanger in an aqueous medium below pH 8.
  • Suitable for extraction of phenols and phenolic pigments, petroleum fractionation, saccharides, drugs, and metabolites.
  • Designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-771 Supplier Waters Corp Supplier No. WAT054650
These Amino Propyl (NH2) Vac cartridges contain a moderately polar, silica-based bonded phase with a weakly basic surface.
  • Sorbent can function as a polar sorbent, offering different selectivity for acidic/basic analytes, or as a weak anion exchanger in an aqueous medium below pH 8.
  • Suitable for extraction of phenols and phenolic pigments, petroleum fractionation, saccharides, drugs, and metabolites.
  • Designed for use with vacuum manifolds and automated SPE instruments.