Learn More
Waters Corp Waters Vac Cartridge, Aminopropyl (NH2), 100 mg, 55–105 μm
Catalog No. 50785193 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These cartridges feature a moderately polar, silica-based bonded phase with a weakly basic surface. They are designed for use with vacuum manifolds and automated SPE instruments for various extraction applications.
  • Moderately polar, silica-based bonded phase with a weakly basic surface.
  • Functions as polar sorbent for acidic/basic analytes or weak anion exchanger below pH 8.
  • Extracts phenols, pigments, petroleum, saccharides, and drugs/metabolites.
  • Syringe-barrel format compatible with vacuum manifolds and automated SPE.
  • Poly-foil pouch maintains product integrity and purity.
  • 20 μm frit (HD/UHMW polyethylenes) offers good flow, low extractables, solvent resistance, and depth filtration.
  • Enables easy, spill-free transfer and extraction of solvents and samples.
Catalog No. 50-785-193 Supplier Waters Corp Supplier No. WAT023610
These cartridges feature a moderately polar, silica-based bonded phase with a weakly basic surface. They are designed for use with vacuum manifolds and automated SPE instruments for various extraction applications.
  • Moderately polar, silica-based bonded phase with a weakly basic surface.
  • Functions as polar sorbent for acidic/basic analytes or weak anion exchanger below pH 8.
  • Extracts phenols, pigments, petroleum, saccharides, and drugs/metabolites.
  • Syringe-barrel format compatible with vacuum manifolds and automated SPE.
  • Poly-foil pouch maintains product integrity and purity.
  • 20 μm frit (HD/UHMW polyethylenes) offers good flow, low extractables, solvent resistance, and depth filtration.
  • Enables easy, spill-free transfer and extraction of solvents and samples.