Learn More
Waters Corp Waters Sep-Pak Aminopropyl (NH2) Vac Cartridge, 20 cc, 5 g
Catalog No. 50785780 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Moderately polar, silica-based bonded phase with weakly basic surface.
  • Can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanger in aqueous medium below pH 8.
  • Applications include extraction of phenols and phenolic pigments, petroleum fractionation, saccharides and drugs and metabolites.
  • Syringe-barrel-type cartridges designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-780 Supplier Waters Corp Supplier No. WAT054695
  • Moderately polar, silica-based bonded phase with weakly basic surface.
  • Can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanger in aqueous medium below pH 8.
  • Applications include extraction of phenols and phenolic pigments, petroleum fractionation, saccharides and drugs and metabolites.
  • Syringe-barrel-type cartridges designed for use with vacuum manifolds and automated SPE instruments.