Learn More
Waters Corp Waters Vac Cartridge, Aminopropyl (NH2), 6 cc, 500 mg Sorbent, 55-105 μm
Catalog No. 50785753 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Vac cartridges feature a moderately polar, silica-based bonded phase with a weakly basic surface. The sorbent can be used as a polar sorbent for acidic/basic analytes or as a weak anion exchanger. Designed for vacuum manifolds and automated SPE instruments, applications include extraction of phenols, petroleum fractionation, saccharides, and drugs.
  • Facilitates easy extraction and transfer of solvents, solutions, and samples, preventing spillage or loss.
  • Acts as a precision chromatographic tool for sample enrichment and purification.
  • Features high-quality packing materials that are clean, dry, and reproducible with optimal characteristics.
  • Product integrity ensured by delivery sealed in a non-contaminating poly-foil pouch.
  • Equipped with a 20 μm frit, offering excellent solvent resistance, low extractables, and depth filtration.
  • Functions optimally within a 2 to 8 pH range.
Catalog No. 50-785-753 Supplier Waters Corp Supplier No. WAT054560
These Vac cartridges feature a moderately polar, silica-based bonded phase with a weakly basic surface. The sorbent can be used as a polar sorbent for acidic/basic analytes or as a weak anion exchanger. Designed for vacuum manifolds and automated SPE instruments, applications include extraction of phenols, petroleum fractionation, saccharides, and drugs.
  • Facilitates easy extraction and transfer of solvents, solutions, and samples, preventing spillage or loss.
  • Acts as a precision chromatographic tool for sample enrichment and purification.
  • Features high-quality packing materials that are clean, dry, and reproducible with optimal characteristics.
  • Product integrity ensured by delivery sealed in a non-contaminating poly-foil pouch.
  • Equipped with a 20 μm frit, offering excellent solvent resistance, low extractables, and depth filtration.
  • Functions optimally within a 2 to 8 pH range.