Learn More
Waters Corp Sep-Pak Vac Cartridge, PS2, 3 cc, 200 mg Sorbent, 80 μm
Catalog No. 50466720 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak PS2 Vac Cartridge is a solid phase extraction product utilizing a styrene-divinylbenzene copolymer. It is suitable for concentrating pesticides and herbicides from water samples.
  • Styrene-divinylbenzene copolymer packed in a short body cartridge.
  • Applications include concentration of pesticides and herbicides in water.
  • Used in Japan pesticide methods #5, #6, and #9.
  • AC2 connected in series with PS2 was adopted as the official method for the extraction of pesticides from water by JPMHW.
Catalog No. 50-466-720 Supplier Waters Corp Supplier No. JJAN20151
The Sep-Pak PS2 Vac Cartridge is a solid phase extraction product utilizing a styrene-divinylbenzene copolymer. It is suitable for concentrating pesticides and herbicides from water samples.
  • Styrene-divinylbenzene copolymer packed in a short body cartridge.
  • Applications include concentration of pesticides and herbicides in water.
  • Used in Japan pesticide methods #5, #6, and #9.
  • AC2 connected in series with PS2 was adopted as the official method for the extraction of pesticides from water by JPMHW.