Learn More
Waters Corp Sep-Pak Cartridge, PSA, 6cc, 500 mg, 30/pk
Catalog No. 502391611 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These solid-phase extraction cartridges offer a quick, repeatable, and disposable solution for sample preparation in analytical labs. The silica-based sorbent, featuring primary and secondary amines, provides high ion-exchange capacity and strong affinity for fatty acids, polar pigments, and sugars.
  • Manufactured in an ISO 13485 and ISO 9002-registered facility, adhering to cGMP standards for Class 1 Medical Devices.
  • Designed for quick, repeatable, and disposable solid-phase extraction.
  • Utilizes a silica-based bonded phase with primary and secondary amines.
  • Provides high ion-exchange capacity and similar selectivity to aminopropyl.
  • Effective for extracting fatty acids, polar pigments, and sugars.
  • Supports both manual and automatic solid-phase extraction applications.
Catalog No. 50-239-1611 Supplier Waters Corp Supplier No. 186004563
These solid-phase extraction cartridges offer a quick, repeatable, and disposable solution for sample preparation in analytical labs. The silica-based sorbent, featuring primary and secondary amines, provides high ion-exchange capacity and strong affinity for fatty acids, polar pigments, and sugars.
  • Manufactured in an ISO 13485 and ISO 9002-registered facility, adhering to cGMP standards for Class 1 Medical Devices.
  • Designed for quick, repeatable, and disposable solid-phase extraction.
  • Utilizes a silica-based bonded phase with primary and secondary amines.
  • Provides high ion-exchange capacity and similar selectivity to aminopropyl.
  • Effective for extracting fatty acids, polar pigments, and sugars.
  • Supports both manual and automatic solid-phase extraction applications.