Learn More
Waters Corp Sep-Pak Accell Plus QMA Vac RC Cartridge, 100 mg Sorbent per Cartridge, 37 - 55 µm, 50/pk
Catalog No. 50785518 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Accell Plus QMA Vac RC cartridges contain a silica-based, hydrophilic, strong anion-exchanger with large pore size (300Å). The cartridges are designed for extraction of anionic analytes in aqueous and non-aqueous solutions. The large pore size makes it ideal for isolation of larger molecules such as anionic proteins such as immunoglobulins and enzymes. Other applications include isolation of phenolic compounds and removal of acidic pigments in wine, fruit juices, and food extracts. These cartridges feature an integral 20 cc funnel-shaped reservoir for convenient processing of larger volumes of samples. They can be used with robotic sample preparation equipment or with vacuum manifolds.
Catalog No. 50-785-518 Supplier Waters Corp Supplier No. WAT043460
Sep-Pak Accell Plus QMA Vac RC cartridges contain a silica-based, hydrophilic, strong anion-exchanger with large pore size (300Å). The cartridges are designed for extraction of anionic analytes in aqueous and non-aqueous solutions. The large pore size makes it ideal for isolation of larger molecules such as anionic proteins such as immunoglobulins and enzymes. Other applications include isolation of phenolic compounds and removal of acidic pigments in wine, fruit juices, and food extracts. These cartridges feature an integral 20 cc funnel-shaped reservoir for convenient processing of larger volumes of samples. They can be used with robotic sample preparation equipment or with vacuum manifolds.