Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Sep-Pak C18 Vac RC Cartridge, 100 mg Sorbent, 55 to 105 μm

Catalog No. 50785413 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These cartridges feature a silica-based bonded phase with strong hydrophobicity, designed to adsorb analytes, even those with weak hydrophobicity, from aqueous solutions. They are suitable for various applications, including the analysis of drugs and their metabolites in biological samples, desalting peptides, detecting trace organics in environmental water, and analyzing organic acids in beverages. Their behavior is similar to reversed-phase HPLC columns.

  • C18 chemistry
  • Endcapped
  • Vac RC Cartridge format
  • Mass spectrometry compatible
  • Reversed phase mode
  • Particle size: 55 to 105 μm
  • Pore size: 125 Å
  • Silica sorbent substrate
  • Sorbent weight: 100 mg
  • Not water wettable
  • pH range: 2 to 8 pH
  • Hold up volume: 0.2 mL

Catalog No. 50-785-413 Supplier Waters Corp Supplier No. WAT036935
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These cartridges feature a silica-based bonded phase with strong hydrophobicity, designed to adsorb analytes, even those with weak hydrophobicity, from aqueous solutions. They are suitable for various applications, including the analysis of drugs and their metabolites in biological samples, desalting peptides, detecting trace organics in environmental water, and analyzing organic acids in beverages. Their behavior is similar to reversed-phase HPLC columns.

  • C18 chemistry
  • Endcapped
  • Vac RC Cartridge format
  • Mass spectrometry compatible
  • Reversed phase mode
  • Particle size: 55 to 105 μm
  • Pore size: 125 Å
  • Silica sorbent substrate
  • Sorbent weight: 100 mg
  • Not water wettable
  • pH range: 2 to 8 pH
  • Hold up volume: 0.2 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.