Learn More
Waters Corp Waters Sep-Pak C18 Vac RC Cartridge, 3 cc, 500 mg, 55–105 μm
Catalog No. 50785415 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge uses a silica-based bonded phase with strong hydrophobicity to adsorb analytes from aqueous solutions, behaving like reversed-phase HPLC columns. It is a general-purpose SPE phase for both polar and non-polar analytes.
  • Silica-based bonded phase, strong hydrophobicity
  • Adsorbs analytes (even weakly hydrophobic) from aqueous solutions
  • Behaves like reversed-phase HPLC columns
  • Offers unique selectivity and excellent recovery of analytes
  • General-purpose SPE phase for polar and non-polar analytes
  • Syringe-barrel cartridge design for vacuum manifolds and automated SPE
  • Manufactured to ISO 9002, ISO 13485, and cGMP standards
  • Convenient, reproducible, and disposable solid-phase extraction device
  • Clean, dry, and reproducible packings in activity and capacity
Catalog No. 50-785-415 Supplier Waters Corp Supplier No. WAT036945
This cartridge uses a silica-based bonded phase with strong hydrophobicity to adsorb analytes from aqueous solutions, behaving like reversed-phase HPLC columns. It is a general-purpose SPE phase for both polar and non-polar analytes.
  • Silica-based bonded phase, strong hydrophobicity
  • Adsorbs analytes (even weakly hydrophobic) from aqueous solutions
  • Behaves like reversed-phase HPLC columns
  • Offers unique selectivity and excellent recovery of analytes
  • General-purpose SPE phase for polar and non-polar analytes
  • Syringe-barrel cartridge design for vacuum manifolds and automated SPE
  • Manufactured to ISO 9002, ISO 13485, and cGMP standards
  • Convenient, reproducible, and disposable solid-phase extraction device
  • Clean, dry, and reproducible packings in activity and capacity