Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp SPE Cartridge, Diol Sorbent, 500 mg, 1.0 mL

Catalog No. 50785745 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This SPE cartridge offers a silica-based Diol polar bonded phase for chromatography.

  • Diol polar phase: alternative to silica (normal-phase), weakly interacting (aqueous).
  • Extracts proteins, peptides, and cosmetic antibiotics.
  • 500 mg sorbent; 20 cc funnel for larger samples.
  • Compatible with manifolds, automated equipment, robotic/manual use.
  • Reversible flow enhances trace enrichment.
  • High quality packings ensure reproducible activity/capacity.
  • HDPE body: excellent solvent resistance, low UV-extractables.
  • Packing weight controlled (±5%) via patented Radial Compression Technology.
  • 20 µm frits: solvent resistant, low extractables, filter debris.
  • Sealed in poly-foil pouch for integrity and purity.

Catalog No. 50-785-745 Supplier Waters Corp Supplier No. WAT054520
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This SPE cartridge offers a silica-based Diol polar bonded phase for chromatography.

  • Diol polar phase: alternative to silica (normal-phase), weakly interacting (aqueous).
  • Extracts proteins, peptides, and cosmetic antibiotics.
  • 500 mg sorbent; 20 cc funnel for larger samples.
  • Compatible with manifolds, automated equipment, robotic/manual use.
  • Reversible flow enhances trace enrichment.
  • High quality packings ensure reproducible activity/capacity.
  • HDPE body: excellent solvent resistance, low UV-extractables.
  • Packing weight controlled (±5%) via patented Radial Compression Technology.
  • 20 µm frits: solvent resistant, low extractables, filter debris.
  • Sealed in poly-foil pouch for integrity and purity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.