Learn More
Waters Corp Sep-Pak Florisil 3 cc Vac RC Cartridge, 500 mg Sorbent per Cartridge, 50 - 200 µm, 50/pk
Catalog No. 50785517 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Polar, highly active, weakly basic sorbent for adsorption of low to moderate polarity species from nonaqueous solutions.
Catalog No. 50-785-517 Supplier Waters Corp Supplier No. WAT043435
Polar, highly active, weakly basic sorbent for adsorption of low to moderate polarity species from nonaqueous solutions.