Learn More
Waters Corp Sep-Pak Vac RC Cartridge, Aminopropyl (NH2), 20 cc, 500 mg, 55 - 105 μm
Catalog No. 50785744 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Amino Propyl (NH2) Vac RC cartridges offer a moderately polar, silica-based bonded phase with a weakly basic surface. They can function as a polar sorbent for acidic/basic analytes or as a weak anion exchanger (below pH 8). Applications include extraction of phenols, saccharides, and drugs. These cartridges have an integral 20 cc funnel-shaped reservoir for larger samples and are compatible with robotic systems or vacuum manifolds.
  • Moderately polar, silica-based bonded phase with weakly basic surface.
  • Can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanger in aqueous medium below pH 8.
  • Integral 20 cc funnel-shaped reservoir for convenient processing of larger volumes of samples.
  • Can be used with robotic sample preparation equipment or with vacuum manifolds.
Catalog No. 50-785-744 Supplier Waters Corp Supplier No. WAT054515
Sep-Pak Amino Propyl (NH2) Vac RC cartridges offer a moderately polar, silica-based bonded phase with a weakly basic surface. They can function as a polar sorbent for acidic/basic analytes or as a weak anion exchanger (below pH 8). Applications include extraction of phenols, saccharides, and drugs. These cartridges have an integral 20 cc funnel-shaped reservoir for larger samples and are compatible with robotic systems or vacuum manifolds.
  • Moderately polar, silica-based bonded phase with weakly basic surface.
  • Can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanger in aqueous medium below pH 8.
  • Integral 20 cc funnel-shaped reservoir for convenient processing of larger volumes of samples.
  • Can be used with robotic sample preparation equipment or with vacuum manifolds.