Learn More
Waters Corp Sep-Pak Aminopropyl (NH2) Vac RC Cartridge, 100 mg Sorbent per Cartridge, 55 - 105 µm, 50/pk
Catalog No. 50785519 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Amino Propyl (NH2) Vac RC cartridges contain a moderately polar, silica-based bonded phase with weakly basic surface. The sorbent can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanges in aqueous medium below pH 8. Applications include extraction of phenols and phenolic pigments, petroleum fractionation, saccharides and drugs and metabolites. These cartridges feature an integral 20 cc funnel-shaped reservoir for convenient processing of larger volumes of samples. They can be used with robotic sample preparation equipment or with vacuum manifolds.
Catalog No. 50-785-519 Supplier Waters Corp Supplier No. WAT043475
Sep-Pak Amino Propyl (NH2) Vac RC cartridges contain a moderately polar, silica-based bonded phase with weakly basic surface. The sorbent can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanges in aqueous medium below pH 8. Applications include extraction of phenols and phenolic pigments, petroleum fractionation, saccharides and drugs and metabolites. These cartridges feature an integral 20 cc funnel-shaped reservoir for convenient processing of larger volumes of samples. They can be used with robotic sample preparation equipment or with vacuum manifolds.