Learn More
Waters Corp Waters Corporation Sep-Pak Cartridge, tC18, 100 mg Sorbent, 37 - 55 μm, Vac RC
Catalog No. 50785513 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Silica-based bonded phase with strong hydrophobicity; trifunctional bonding chemistry gives it an increased hydrolytic stability; applications similar to C18.
  • Silica-based bonded phase with strong hydrophobicity
  • Trifunctional bonding chemistry for increased hydrolytic stability
  • Applications similar to C18
Catalog No. 50-785-513 Supplier Waters Corp Supplier No. WAT043410
Silica-based bonded phase with strong hydrophobicity; trifunctional bonding chemistry gives it an increased hydrolytic stability; applications similar to C18.
  • Silica-based bonded phase with strong hydrophobicity
  • Trifunctional bonding chemistry for increased hydrolytic stability
  • Applications similar to C18