Learn More
Waters Corp Waters Vac Cartridge, Silica, 1 g Sorbent, 55 to 105 μm
Catalog No. 50785408 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polar stationary phase adsorbs analytes from non-polar solvents (hydrocarbons, chloro- or fluoro-substituted hydrocarbons, esters, ethers). Analyte retention occurs via hydrogen bonding, dipole-dipole interactions, or cation-exchange in aqueous samples. This convenient, reproducible, disposable solid-phase extraction (SPE) device meets ISO 9002, ISO 13485, and cGMP standards.
  • Polar stationary phase.
  • Primarily used to adsorb analytes from non-polar solvents such as hydrocarbons, chloro- or fluoro-substituted hydrocarbons, ethers, or less polar esters.
  • Analyte retention through hydrogen bonding, dipole-dipole interactions, or cation exchange in aqueous samples.
  • Manufactured to ISO 9002, ISO 13485, and cGMP standards.
  • Convenient, reproducible, and disposable SPE device.
  • Ships with 30 cartridges per package.
Catalog No. 50-785-408 Supplier Waters Corp Supplier No. WAT036910
This polar stationary phase adsorbs analytes from non-polar solvents (hydrocarbons, chloro- or fluoro-substituted hydrocarbons, esters, ethers). Analyte retention occurs via hydrogen bonding, dipole-dipole interactions, or cation-exchange in aqueous samples. This convenient, reproducible, disposable solid-phase extraction (SPE) device meets ISO 9002, ISO 13485, and cGMP standards.
  • Polar stationary phase.
  • Primarily used to adsorb analytes from non-polar solvents such as hydrocarbons, chloro- or fluoro-substituted hydrocarbons, ethers, or less polar esters.
  • Analyte retention through hydrogen bonding, dipole-dipole interactions, or cation exchange in aqueous samples.
  • Manufactured to ISO 9002, ISO 13485, and cGMP standards.
  • Convenient, reproducible, and disposable SPE device.
  • Ships with 30 cartridges per package.