Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Silica, 500 mg Sorbent, 3 cc, 55–105 μm
Catalog No. 50785111 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak Silica 3 cc Vac Cartridge is designed for sample preparation, utilizing a polar stationary phase to adsorb analytes from non-polar solvents. It ensures high-quality extraction and purification with a pH range of 1–8 pH.
  • Highest quality packings specifically designed for sample preparation.
  • Packings are clean, dry, and offer repeatable activity and capacity.
  • Provides excellent surface area, pore size, and particle size distributions.
  • Product integrity maintained throughout manufacturing and delivery.
  • Sealed in a non-contaminating poly-foil bag to preserve purity and inactivity.
  • Manufactured to cGMP, ISO 9001 certified facility standards.
  • Operating pH range of 1–8 pH.
  • Ideal for sample enrichment and purification as a precision chromatographic instrument.
Catalog No. 50-785-111 Supplier Waters Corp Supplier No. WAT020810
The Sep-Pak Silica 3 cc Vac Cartridge is designed for sample preparation, utilizing a polar stationary phase to adsorb analytes from non-polar solvents. It ensures high-quality extraction and purification with a pH range of 1–8 pH.
  • Highest quality packings specifically designed for sample preparation.
  • Packings are clean, dry, and offer repeatable activity and capacity.
  • Provides excellent surface area, pore size, and particle size distributions.
  • Product integrity maintained throughout manufacturing and delivery.
  • Sealed in a non-contaminating poly-foil bag to preserve purity and inactivity.
  • Manufactured to cGMP, ISO 9001 certified facility standards.
  • Operating pH range of 1–8 pH.
  • Ideal for sample enrichment and purification as a precision chromatographic instrument.