Learn More
Waters Corp Sep-Pak Vac Cartridge, Silica, 500 mg, 6 cc, 55-105 μm
Catalog No. 50785511 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge utilizes a polar stationary phase, primarily for adsorbing analytes from non-polar solvents like hydrocarbons, chloro- or fluoro-substituted hydrocarbons, or less polar esters and ethers. Analyte retention can occur via hydrogen bonding, dipole-dipole interactions, or cation-exchange in aqueous samples.
  • Designed as a precision chromatographic tool for sample enrichment and purification.
  • Contains a polar stationary phase for adsorbing analytes from non-polar solvents.
  • Manufactured with high-quality, clean, dry, and reproducible packings.
  • Packings offer optimal surface area, pore-size, and particle-size distributions.
  • Sealed in a non-contaminating poly-foil pouch to maintain integrity and purity until opened.
  • Enables easy sample extraction and preparation and convenient transfer without spillage or evaporation.
Catalog No. 50-785-511 Supplier Waters Corp Supplier No. WAT043400
This cartridge utilizes a polar stationary phase, primarily for adsorbing analytes from non-polar solvents like hydrocarbons, chloro- or fluoro-substituted hydrocarbons, or less polar esters and ethers. Analyte retention can occur via hydrogen bonding, dipole-dipole interactions, or cation-exchange in aqueous samples.
  • Designed as a precision chromatographic tool for sample enrichment and purification.
  • Contains a polar stationary phase for adsorbing analytes from non-polar solvents.
  • Manufactured with high-quality, clean, dry, and reproducible packings.
  • Packings offer optimal surface area, pore-size, and particle-size distributions.
  • Sealed in a non-contaminating poly-foil pouch to maintain integrity and purity until opened.
  • Enables easy sample extraction and preparation and convenient transfer without spillage or evaporation.