Learn More
Waters Corp Sep-Pak Silica 3 cc Vac Cartridge, 200 mg Sorbent per Cartridge, 55 - 105 µm, 50/pk
Catalog No. 50785820 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
A polar stationary phase, used primarily to adsorb analytes from non-polar solvents like hydrocarbons, chloro- or fluoro-substituted hydrocarbons or less polar esters and ethers. Analyte retention can occur through hydrogen bonding, dipole-dipole interactions or in aqueous samples through cation-exchange.
Catalog No. 50-785-820 Supplier Waters Corp Supplier No. WAT054930
A polar stationary phase, used primarily to adsorb analytes from non-polar solvents like hydrocarbons, chloro- or fluoro-substituted hydrocarbons or less polar esters and ethers. Analyte retention can occur through hydrogen bonding, dipole-dipole interactions or in aqueous samples through cation-exchange.