Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Silica, 35 cc, 10 g Sorbent, 55–105 μm
Catalog No. 50785502 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polar sorbent simplifies sample extraction and preparation, enabling quick and efficient transfer of solvents and samples without spillage or loss. It functions as a precision chromatographic tool for enrichment and purification.
  • Simplifies sample extraction and preparation, ensuring quick and efficient transfer.
  • Features high-quality packings with good surface area, pore size, and particle size distributions.
  • Manufactured in a cGMP, ISO 9001 certified facility.
Catalog No. 50-785-502 Supplier Waters Corp Supplier No. WAT043355
This polar sorbent simplifies sample extraction and preparation, enabling quick and efficient transfer of solvents and samples without spillage or loss. It functions as a precision chromatographic tool for enrichment and purification.
  • Simplifies sample extraction and preparation, ensuring quick and efficient transfer.
  • Features high-quality packings with good surface area, pore size, and particle size distributions.
  • Manufactured in a cGMP, ISO 9001 certified facility.