Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Silica, 20 cc, 5 g, 55-105 μm
Catalog No. 50785412 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica vac cartridge is a polar sorbent used to adsorb analytes from non-polar solvents. It is a chromatographic tool for sample enrichment and purification, featuring high-quality, consistent packings. Its non-contaminating poly-foil packet ensures integrity until opened. A 20 μm frit, made of HD and UHMW polyethylenes, resists solvents, has low extractables, high flow, and acts as a depth filter. Manufactured in a cGMP, ISO 9001 facility, it allows effortless extraction and transport without spillage or evaporation.
  • Polar sorbent for non-polar solvent analyte adsorption.
  • High-quality, consistent packings.
  • Non-contaminating poly-foil packaging.
  • 20 μm frit: solvent-resistant, low extractables, high flow, depth filter.
  • cGMP, ISO 9001 certified manufacturing.
  • Effortless extraction and transport without spillage.
  • Compatible with 1-8 pH range.
Catalog No. 50-785-412 Supplier Waters Corp Supplier No. WAT036930
This silica vac cartridge is a polar sorbent used to adsorb analytes from non-polar solvents. It is a chromatographic tool for sample enrichment and purification, featuring high-quality, consistent packings. Its non-contaminating poly-foil packet ensures integrity until opened. A 20 μm frit, made of HD and UHMW polyethylenes, resists solvents, has low extractables, high flow, and acts as a depth filter. Manufactured in a cGMP, ISO 9001 facility, it allows effortless extraction and transport without spillage or evaporation.
  • Polar sorbent for non-polar solvent analyte adsorption.
  • High-quality, consistent packings.
  • Non-contaminating poly-foil packaging.
  • 20 μm frit: solvent-resistant, low extractables, high flow, depth filter.
  • cGMP, ISO 9001 certified manufacturing.
  • Effortless extraction and transport without spillage.
  • Compatible with 1-8 pH range.