Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, tC18, 5 g Sorbent, 37-55 μm
Catalog No. 50785504 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sep-Pak Vac Cartridge features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry, providing increased hydrolytic stability. Its applications are similar to C18 cartridges.
  • Silica-based bonded phase with strong hydrophobicity.
  • Trifunctional bonding chemistry provides increased hydrolytic stability.
  • Applications are similar to C18.
  • Mass spec compatible.
  • Endcapped.
  • Reversed phase mode.
Catalog No. 50-785-504 Supplier Waters Corp Supplier No. WAT043365
This Sep-Pak Vac Cartridge features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry, providing increased hydrolytic stability. Its applications are similar to C18 cartridges.
  • Silica-based bonded phase with strong hydrophobicity.
  • Trifunctional bonding chemistry provides increased hydrolytic stability.
  • Applications are similar to C18.
  • Mass spec compatible.
  • Endcapped.
  • Reversed phase mode.