Learn More
Waters Corp Sep-Pak Vac Cartridge, tC18, 200 mg, 37 to 55 μm
Catalog No. 50785819 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak tC18 Vac Cartridge features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry for increased hydrolytic stability. Designed for analyte isolation and enrichment from biological, environmental, and food samples, it also facilitates solvent switching and removal of sample components that elute after analytes of interest on liquid chromatography columns.
  • Streamlines protocols and simplifies sample handling.
  • Eliminates emulsion challenges.
  • Reduces exposure to dangerous agents, improving safety.
  • Allows direct field sampling, bypassing lab transfer.
  • Packing material secured by two polyethylene frits.
  • Cartridge body made of molded, high-purity polypropylene.
  • Connects to multi-position vacuum manifolds via male-Luer tip.
  • Constructed from inert polypropylene to prevent analyte reactions.
Catalog No. 50-785-819 Supplier Waters Corp Supplier No. WAT054925
The Sep-Pak tC18 Vac Cartridge features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry for increased hydrolytic stability. Designed for analyte isolation and enrichment from biological, environmental, and food samples, it also facilitates solvent switching and removal of sample components that elute after analytes of interest on liquid chromatography columns.
  • Streamlines protocols and simplifies sample handling.
  • Eliminates emulsion challenges.
  • Reduces exposure to dangerous agents, improving safety.
  • Allows direct field sampling, bypassing lab transfer.
  • Packing material secured by two polyethylene frits.
  • Cartridge body made of molded, high-purity polypropylene.
  • Connects to multi-position vacuum manifolds via male-Luer tip.
  • Constructed from inert polypropylene to prevent analyte reactions.