Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, tC18, 3 cc, 500 mg Sorbent
Catalog No. 50785400 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid phase extraction cartridge features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry, enhancing hydrolytic stability. It is designed for analyte isolation and enrichment from biological, environmental, and dietary samples, and is compatible with multi-position vacuum manifolds.
  • Features a silica-based bonded phase with strong hydrophobicity.
  • Offers trifunctional bonding chemistry for increased hydrolytic stability.
  • Ideal for analyte isolation and enrichment from biological, environmental, and food samples.
  • Simplifies protocols, minimizes sample handling, and helps avoid emulsion issues.
  • Enhances safety by reducing exposure to hazardous agents and facilitating disposal.
  • Supports direct field sampling, removing the need for sample transport to a lab.
  • Constructed from high-grade, highly inert polypropylene.
Catalog No. 50-785-400 Supplier Waters Corp Supplier No. WAT036815
This solid phase extraction cartridge features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry, enhancing hydrolytic stability. It is designed for analyte isolation and enrichment from biological, environmental, and dietary samples, and is compatible with multi-position vacuum manifolds.
  • Features a silica-based bonded phase with strong hydrophobicity.
  • Offers trifunctional bonding chemistry for increased hydrolytic stability.
  • Ideal for analyte isolation and enrichment from biological, environmental, and food samples.
  • Simplifies protocols, minimizes sample handling, and helps avoid emulsion issues.
  • Enhances safety by reducing exposure to hazardous agents and facilitating disposal.
  • Supports direct field sampling, removing the need for sample transport to a lab.
  • Constructed from high-grade, highly inert polypropylene.