Learn More
Waters Corp Waters Sep-Pak tC18 Vac Cartridge, 6 cc, 1 g Sorbent
Catalog No. 50785396 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak tC18 6 cc Vac Cartridge features a silica-based bonded phase with strong hydrophobicity and a trifunctional bonding chemistry for increased hydrolytic stability. These cartridges are designed for analyte isolation and enrichment from biological, environmental, and food samples. They are suitable for use in the pH range of 2 to 8, with a hold-up volume of 1.9 mL and 1 g sorbent.
  • Silica-based bonded phase with strong hydrophobicity.
  • Trifunctional bonding chemistry for increased hydrolytic stability.
  • Designed for analyte isolation and enrichment from biological, environmental, and food samples.
  • Operates in the pH range of 2 to 8.
  • Supports a hold-up volume of 1.9 mL and 1 g sorbent.
  • Available in various designs for manual and automatic solid-phase extraction.
Catalog No. 50-785-396 Supplier Waters Corp Supplier No. WAT036795
The Sep-Pak tC18 6 cc Vac Cartridge features a silica-based bonded phase with strong hydrophobicity and a trifunctional bonding chemistry for increased hydrolytic stability. These cartridges are designed for analyte isolation and enrichment from biological, environmental, and food samples. They are suitable for use in the pH range of 2 to 8, with a hold-up volume of 1.9 mL and 1 g sorbent.
  • Silica-based bonded phase with strong hydrophobicity.
  • Trifunctional bonding chemistry for increased hydrolytic stability.
  • Designed for analyte isolation and enrichment from biological, environmental, and food samples.
  • Operates in the pH range of 2 to 8.
  • Supports a hold-up volume of 1.9 mL and 1 g sorbent.
  • Available in various designs for manual and automatic solid-phase extraction.