Learn More
Waters Corp Waters Vac Cartridge, tC2, 1 cc, 100 mg Sorbent, 37–55 μm
Catalog No. 50785660 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based bonded phase cartridge features low hydrophobicity and is designed for methods requiring less retention than C8, with applications similar to C18 and C8.
  • Low hydrophobicity character
  • Suitable for methods requiring less retention than C8
  • Applications are similar to C18 and C8
Catalog No. 50-785-660 Supplier Waters Corp Supplier No. WAT052710
This silica-based bonded phase cartridge features low hydrophobicity and is designed for methods requiring less retention than C8, with applications similar to C18 and C8.
  • Low hydrophobicity character
  • Suitable for methods requiring less retention than C8
  • Applications are similar to C18 and C8