Learn More
Waters Corp Sep-Pak Vac Cartridge, tC2, 3 cc, 500 mg, 37–55 μm
Catalog No. 50785661 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Vac Cartridge utilizes a silica-based bonded phase with low hydrophobicity character. It is designed for methods requiring less retention than C8, and its applications are similar to those of C18 and C8 phases.
  • Silica-based bonded phase with low hydrophobicity character.
  • Use for methods requiring less retention than C8.
  • Applications are similar to C18 and C8.
Catalog No. 50-785-661 Supplier Waters Corp Supplier No. WAT052715
This Vac Cartridge utilizes a silica-based bonded phase with low hydrophobicity character. It is designed for methods requiring less retention than C8, and its applications are similar to those of C18 and C8 phases.
  • Silica-based bonded phase with low hydrophobicity character.
  • Use for methods requiring less retention than C8.
  • Applications are similar to C18 and C8.