Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Sepabeads™ SP-207

Catalog No. 111001829 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1829 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1829 Supplier MilliporeSigma™ Supelco™ Supplier No. 13623U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Swelling in toluene is 18%

Sepabeads™ SP-207 is a nonionic polystyrene-divinylbenzene polymer resin used as column packing material. It is widely used as a adsorbent for purification in biopharmaceutical applications.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bucket
Quantity 1000 g
Surface Area (Metric) 650 m2/g
Product Line Sepabeads
Density 1.18 g/mL at 25°C (true wet)(lit.)
CAS 905310-11-4

Sepabeads is a registered trademark of Mitsubishi Chemical Corp.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.